Order Entry
Slovenia
ContactUsLinkComponent
Human recombinant ZG16 (from cells)
Human recombinant ZG16 (from cells)
Catalog # PRSI91-556
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant ZG16 (from cells)
Catalog # PRSI91-556
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Zymogen Granule Membrane Protein 16|hZG16|Secretory Lectin ZG16|Zymogen Granule Protein 16|ZG16
  • Protein/peptide name:
    ZG16
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    17.7 kD
  • Sequence:
    Asn17-Cys167
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Zymogen Granule Membrane Protein 16 (ZG16) belongs to the jacalin lectin family. ZG16 is highly expressed in liver and is detected at lower levels in colon, ileum and jejunum. ZG16 may play a role in protein trafficking. In addition, ZG16 may act as a linker molecule between the submembranous matrix on the luminal side of zymogen granule membrane (ZGM) and aggregated secretory proteins during granule formation in the TGN.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag