Order Entry
Slovenia
ContactUsLinkComponent
Human recombinant SP-D (from cells)
Human recombinant SP-D (from cells)
Catalog # PRSI91-491
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant SP-D (from cells)
Catalog # PRSI91-491
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Pulmonary Surfactant-Associated Protein D|Lung Surfactant Protein D|PSPD|COLEC7|SFTP4|SP-D|SFTPD|PSP-D|Collectin-7
  • Protein/peptide name:
    SP-D
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    36.46 kD
  • Sequence:
    Ala21-Phe375(Glu22Gly
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

Surfactant Pulmonary-Associated Protein D (SP-D) is a 43 kDa member of the collectin family of innate immune modulators. Its principal components consist of a collagen-like region and a C-terminal carbohydrate recognition domain (CRD), a structure that places it in a subset of pattern recognition proteins termed defense collagens. SP-D is constitutively secreted by alveolar lining cells and epithelium associated with tubular structures and induced in cardiac smooth muscle and endothelial cells. It binds both secreted and transmembrane proteins that transduce its function. It binds human neutrophil defensins, modulating influenza anti-viral defense. It binds MD-2/LY96, a secreted protein that cooperates with Toll-like receptors (TLRs) in the response of macrophages to bacterial lipopolysaccharides (LPS) or cell wall components. It also binds macrophage CD14 and TLRs directly, blocking binding of LPS and down-regulating TNF- alpha secretion. SP-D binding of both SIRP alpha and the calreticulin/CD91 complex on macrophages allows for a graded response to environmental challenge.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag