Order Entry
Slovenia
ContactUsLinkComponent
Human recombinant serpin E2 (from HEK293 cells)
Human recombinant serpin E2 (from HEK293 cells)
Catalog # PRSI96-683
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant serpin E2 (from HEK293 cells)
Catalog # PRSI96-683
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Biological activity:
    Measured by its ability to inhibit trypsin cleavage of a fluorogenic peptide substrate, Mca - RPKPVE - Nval - WRK(Dnp) - NH2.
  • Protein synonyms:
    Glia-derived nexin|PI7|SERPINE2|GDN|SerpinE2|Peptidase inhibitor 7|PN-1|Serpin E2|PI-7|Protease nexin 1
  • Protein/peptide name:
    Serpin E2
  • Purity:
    >92% as determined by SDS-PAGE.
  • Molecular weight:
    42.7 kDa
  • Formulation:
    Lyophilized, 20 mM NaAc, 100 mM NaCl, PH6.5
  • Tested applications:
    Western Blot

Specifications

About this item

SERPINE2 is also known as Glia-derived nexin (GDN), Peptidase inhibitor 7 (PI7), Protease nexin 1(PN1). SERPINE2 is a secreted glycoprotein which belongs to the serpin family. SerpinE1 is the primary physiological inhibitor of the two plasminogen activators urokinase (uPA) and tissue plasminogen activator (tPA). PAI-1 / GDN is also implicated in adipose tissue development. It suggests that PAI-1 inhibitors serve in the control of atherothrombosis. Defects in Serpin E1 / PN1 are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency) which is characterized by abnormal bleeding due to SerpinE1 defect in the plasma.