Specifications
- Pk:25 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Biological activity:Measured by its binding ability in a functional ELISA. Immobilized Human PD-1, Strep Tag at 2 ug/mL ( 100 ul/well ) can bind Human PD-L2, Fc Tag with a linear range of 0.4-3 ng/mL. Measured by its binding ability in a functional ELISA. Immobilized Human PD-1, Strep Tag at 5 ug/mL ( 100 ul/well ) can bind Human PD-L1, Fc Tag (HPLC-verified) with a linear range of 1-50 ng/mL.
- Protein synonyms:PD-1|hPD-1|hPD-l|CD279|PDCD1|PD1|SLEB2
- Protein/peptide name:PD1
- Purity:>95% as determined by SDS-PAGE.
- Molecular weight:19.5 kDa
- Formulation:Lyophilized, PBS, pH7.4
- Tested applications:Western Blot
Specifications
About this item
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.