Human recombinant Bace13 (from HEK293 cells)
Catalog # PRSI96-052
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:200 µG
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Environmentally Preferable:
- Biological activity:Measured by its ability to cleave a fluorogenic peptide substrate, Mca-SEVNLDAEFRK (Dpn) RR-NH2. The specific activity is > 1. 5 pmoles /min /μg.
- Protein synonyms:Memapsin-2|KIAA1149|BACE1|HSPC104|ASP2|FLJ90568|BACE
- Protein/peptide name:Bace13
- Purity:>85% as determined by SDS-PAGE.
- Molecular weight:75 kDa
- Formulation:Lyophilized, 50 mM tris, 100 mM glycine, pH7.5
- Tested applications:Western Blot