Anti-ADCY6 Chicken Polyclonal Antibody
PRSIXW-7717
- Antigen name:Adenylate Cyclase 6
- Antigen symbol:ADCY6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:LCCS8|KIAA0422|AC6
- Concentration:1 mg/mL
- Immunogen:1-121
- Tested applications:Immunoaffinity Purified
- Size:50 µg
- Pk:50 µG
FUNCTION: Membrane-bound, calcium-inhibitable adenylyl cyclase (By similarity).
CATALYTIC ACTIVITY: ATP = 3',5'-cyclic AMP + diphosphate.
SIMILARITY: Contains 2 guanylate cyclase domains.
COFACTOR: Binds 2 magnesium ions per subunit (By similarity).
Adenylate cyclase type VI antibody can be used for the detection of Adenylate cyclase type VI by Western Blot.
Phosphate-Buffered Saline. No preservatives added.
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ADCY6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human