Specifications
- Antigen name:Tumor Necrosis Factor Receptor Superfamily Member 13B
- Antigen symbol:TNFRSF13B
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:CD267|Transmembrane activator and CAML interactor|CVID|RYZN|TACI|Tumor necrosis factor receptor superfamily member 13B|CVID2|TNFRSF14B
- Immunogen:Produced from sera of goats pre-immunized with highly pure (>98%) recombinant hTACI (human TACI).
- Size:50 µg
- Pk:50 µG
Specifications
About this item
TACI is a lymphocyte specific member of the tumor necrosis factor (TNF) receptor superfamily. It interacts with calcium-modulator and cyclophilin ligand (CAML). The protein binds to both APRIL and BAFF and induces activation of the transcription factors NFAT (nuclear-factor of activated T cells), AP1, and NF kappa B and plays a crucial role in humoral immunity by interacting with a TNF ligand. This gene is located within the Smith Magenis syndrome region on chromosome 17.
Anti-hTACI specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TNFRSF13B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human