Specifications
- Antigen name:Tissue inhibitor of metalloproteinases 1
- Antigen symbol:TIMP1
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Metalloproteinase inhibitor 1|CLGI|Fibroblast collagenase inhibitor|Tissue inhibitor of metalloproteinases|TIMP1|TIMP|Collagenase inhibitor|EPA|Erythroid-potentiating activity|TIMP-1
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hTIMP-1 (human Tissue Inhibitor of Metalloproteinases-1). Human TIMP-1 specific antibody was purified by affinity chromatography and then biotinylated.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Tissue inhibitor of metalloproteinases 1 (TIMP1) and Tissue inhibitor of metalloproteinases 2 (TIMP 2) have similar properties, specifically in inhibiting enzymes of the matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP1 forms a complex of 1:1 stoichiometry with activated interstitial collagenases, activated stromelysin, active form of 72kDa Type IV collagenase (also known as MMP2 or gelatinase A), and latent and active forms of 92kDa Type IV collagenase (also known as MMP0 or gelatinase B). TIMPs inhibit the proteolytic invasiveness of tumour cells and normal placental trophoblast cells. TIMP1 is produced in low (pg/mL) levels by most cell types. Treatment of cells with the phorbol ester TPA stimulates production of TIMP1 in some cell types, but the low protein levels produced often require concentration of cell culture media to visualize the bands by Western Blotting.
The reconstituted antibody can temporarily be stored at 2-8°C. Long term storage at -20°C or below. Avoid repeated freeze-thaw cycles.
Type:
Antigen: TIMP1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human