Anti-NXPE4 Rabbit Polyclonal Antibody
PRSI31-375
- Antigen name:Neurexophilin And PC-Esterase Domain Family, Member 4
- Antigen symbol:NXPE4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:C11orf33|FAM55D
- Immunogen:A synthetic peptide corresponding a region of human FAM55D.
- Size:100 µg
- Pk:100 µG
The function remains unknown.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NXPE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Dog