Anti-SLC18A1 Rabbit Polyclonal Antibody
Catalog # PRSI28-326
Supplier: ProSci Inc.
Specifications
- Antigen name:Solute Carrier Family 18 Member A1
- Antigen symbol:SLC18A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human SLC18A1.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SLC18A1 is an integral protein in the membrane of secretory vesicles of neuroendocrine and endocrine cells that allows the transport of biogenic monoamines, such as serotonin, from the cytoplasm into the secretory vesicles.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC18A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human