Specifications
- Antigen name:Kinesin heavy chain member 2A
- Antigen symbol:KIF2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:HK2|KIF2|CDCBM3
- Immunogen:A synthetic peptide corresponding a region of human KIF2A.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
KIF2A belongs to the kinesin-like protein family, MCAK/KIF2 subfamily. It contains 1 kinesin-motor domain. KIF2A plus end-directed microtubule-dependent motor required for normal brain development. It may regulate microtubule dynamics during axonal growth. KIF2A is implicated in formation of bipolar mitotic spindles. It has microtubule depolymerization activity. Hela cells lacking KIF2A show asymmetric or monopolar mitotic spindles. Osteosarcoma cells (U2OS) lacking KIF2A or KIF2B show disorganised or monopolar mitotic spindles.Kinesins, such as KIF2, are microtubule-associated motor proteins. For background information on kinesins, see MIM 148760.
KIF2A antibody can be used for detection of KIF2A by ELISA at 1:62500. KIF2A antibody can be used for detection of KIF2A by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: KIF2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat