Order Entry
Slovenia
ContactUsLinkComponent
Anti-GPAT1 Rabbit Polyclonal Antibody
Anti-GPAT1 Rabbit Polyclonal Antibody
Catalog # PRSI4613
Supplier:  ProSci Inc.
undefined
Anti-GPAT1 Rabbit Polyclonal Antibody
Catalog # PRSI4613
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Glycerol-3-phosphate Acyltransferase 1
  • Antigen symbol:
    GPAT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    GPAT1|GPAT
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    GPAT1 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of the human GPAT1.
  • Tested applications:
    E, WB, IHC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

GPAT1 Antibody: Glycerol-3-phosphate acyltransferase 1 (GPAT1), one of four known GPAT isoforms, is located on the mitochondrial outer membrane, allowing reciprocal regulation with carnitine palmitoyltransferase-1. It is thought to be critical for the development of hepatic steatosis; steatosis triggered by GPAT1 overexpression leads to hepatic and possibly peripheral insulin resistance. GPAT1 is transcriptionally upregulated by insulin and sterol regulatory element binding protein (SREBP-1) and downregulated by AMP-activated protein kinase. Mice deficient in GPAT1 exhibit decreased triacylglycerol (TAG) in cardiomyocytes even in high-fat diets, suggesting that GPAT1 contributes significantly to TAG accumulation in heart tissue during lipogenic or high fat diets. At least two isoforms of GPAT1 are known to exist.

Type:
Antigen: GPAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat