Order Entry
Slovenia
ContactUsLinkComponent
Tizanidine Related Compounds, Reference Standard
Tizanidine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Tizanidine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    64461-82-1
  • Formula:
    C₉H₉Cl₂N₅S
  • Description:
    Tizanidine Hydrochloride
  • Synonym(s):
    4-amino-5-chloro-2,1,3-benzothiadiazole

Specifications

Frequently Bought Together