Order Entry
Slovenia
ContactUsLinkComponent
 
Ticlopidine Related Compounds, Reference Standard
 :  USP
 
Ticlopidine Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    68559-60-4
  • Synonym(s):
    4-oxo-4,5,6,7-tetrahydrothieno-[3,2-c]pyridine
  • Formula:
    C₇H₇NOS
  • Description:
    Ticlopidine Related Compound A

 

Frequently Bought Together