Anti-TNFRSF11A Rabbit Polyclonal Antibody
Catalog # BSBTA01064-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Tumor necrosis factor receptor superfamily member 11A
- Antigen symbol:TNFRSF11A
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 11A(TNFRSF11A) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat