Order Entry
Slovenia
ContactUsLinkComponent
Rabeprazole Related Compounds, Reference Standard
Rabeprazole Related Compounds, Reference Standard
Supplier:  USP
Certificates
Rabeprazole Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    583-39-1
  • UN No.:
    2811
  • Synonym(s):
    1H-Benzimidazole-2-thiol
  • Formula:
    C₇H₆N₂S
  • Description:
    Rabeprazole Related Compound C

Specifications

Frequently Bought Together