beta-1,3-Glucan (from Euglena gracilis)
Supplier: Merck
Certificates
About this item
β-1,3-Glucan from Euglena gracilis may be compared with other β-1,3-glucans and their derivatives to study mechanisms of protection of DNA from oxidative lesions. And are used as a substrate to identify, differentiate and characterise glucanase(s).
Specifications
- Source:Euglena gracilis
- Protein synonyms:Paramylon
- Protein/peptide name:beta-1,3-Glucan
- Molecular weight:Approx 500000 Da
Specifications
Frequently Bought Together