Prostaglandin A1-biotin solution ≥95% in ethanol
:
:C₃₅H₅₈N₄O₅S
:646.9 g/mol
:Freezer
Frequently Bought Together
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR® Blender Bags
: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Premium Line, Serological Pipettes
: VWR Collection
VWR®, Test Tubes without Rim
: VWR Collection
Transfer pipettes
: RATIOLAB
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
VWR®, Spreaders, L-Shaped, Standard Line
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Petri dishes, square
: Greiner Bio-One