Anti-PIP Mouse Monoclonal Antibody [clone: PIP/1571]
ABNOMAB14870
: Abnova
- Antibody type:Primary
- Antigen symbol:PIP
- Clonality:Monoclonal
- Clone:PIP/1571
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:5304
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 41-146 of human PIP.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against partial recombinant human PIP.
Type: Primary
Antigen: PIP
Clonality: Monoclonal
Clone: PIP/1571
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human