(3E)-Testosterone-O-(carboxymethyl)oxime : BSA ≥98% (by TLC), BSA, Sigma-Aldrich®
Catalog # SIALT3392-10MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Seq:2275
Specifications
Specification Test Results
- Appearance (Form):Lyophilised powder
- Appearance (Colour):White
- Solubility (Solvent):Phosphate buffer
- Solubility (pH):7,50 - 7,50
- Solubility (Conc.):1,90 - 2,10 mg/ml
- Solubility (Turbidity):Clear to slightly hazy
- Solubility (Colour):Colourless
- Purity (TLC):≥98,00%
- Conjugation (Ratio Steroid/BSA):20,00 - 30,00 mole/m
- Storage temperature:2 - 8 °C
- Extent of labeling:20 - 30 mol steroid per mol BSA
Specification Test Results
About this item
:
:361.482 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
CAS Number:10190-93-9
Formula:C₂₁H₃₁NO₄
MDL Number:MFCD00132414