Zeolite <45 µm, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1318-02-1
:
Formula:Na₂O·Al₂O₃·xSiO₂·yH₂O
MDL Number:MFCD00132601
Specification Test Results
- Particle size:<45 µm
Specification Test Results
Frequently Bought Together