Zinc citrate dihydrate ≥97%, powder, Sigma-Aldrich®
Catalog # SIAL480762-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance (Color):White to Off-White
- Appearance (Form):Powder
- Complexiometric EDTA (Zn):31,0 - 33,3%
- ICP major component:Confirms zinc component
- Assay:97%
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ100
:610.368 g/mol
CAS Number:5990-32-9
Formula:C₁₂H₁₄O₁₆Zn₃
MDL Number:MFCD00150587