Order Entry
Slovenia
ContactUsLinkComponent
Cell detachment solution, Accumax™
Cell detachment solution, Accumax™
Catalog # SIALA7089-100ML
Supplier:  Merck
CAS Number:  
Cell detachment solution, Accumax™
Catalog # SIALA7089-100ML
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Accumax™ cell detachment solution
Specifications
About this item
Accumax™ is a cell detachment solution consisting of proteolytic and collagenolytic enzymes.
  • Sterile-filtered
  • Suitable for cell culture
Accumax has proven effective in dissociating clumped cells in suspension cultures of hybridomas, CHO, BHK, 293, COS and Sf9 cells. The solution performed extremely well in dissociating cell counting, viral transfection assays, cell sorting, flow cytometry, and bioreactor scale-up. It has also been used to create single cell suspensions from clumped cell cultures for accurate cell counting, detachment of cells from primary tissue, and the routine detachment of cells from standard tissue culture plasticware and adhesion coated plasticware.