Anti-COMMD3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-8183R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:COMM Domain Containing 3
- Antigen symbol:COMMD3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23412
- Antigen synonyms:C10orf8 COMM domain containing protein 3|BUP|Protein PIL|COMD3_HUMAN.|FLJ45471|DKFZp686K0399
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
COMMD3 is widely expressed, with highest expression in thymus.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: COMMD3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: