Anti-ANP32E Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-6862R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Acidic Nuclear Phosphoprotein 32 Family, Member E
- Antigen symbol:ANP32E
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:81611
- Antigen synonyms:LANPL|LANP-L
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Inhibits activity of protein phosphatase 2A (PP2A). Does not inhibit protein phosphatase 1. May play a role in cerebellar development and synaptogenesis process by modulating PP2A activity.Tissue specificityExpressed in peripheral blood leukocytes, colon, small intestine, prostate, thymus, spleen, skeletal muscle, liver and kidney.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ANP32E
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat