Order Entry
Slovenia
ContactUsLinkComponent
Anti-SLC29A4 Rabbit Polyclonal Antibody (Cy5®)
Anti-SLC29A4 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-4176R-CY5
Supplier:  Bioss
Anti-SLC29A4 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-4176R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 29 Member A4
  • Antigen symbol:
    SLC29A4
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    222962
  • Antigen synonyms:
    ENT4|PSEC0113|Solute carrier family 29 member 4|hENT4|Plasma membrane monoamine transporter|Equilibrative nucleoside transporter 4|PMAT|SLC29A4
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    440-490/530
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Functions as a polyspecific organic cation transporter, efficiently transporting many organic cations such as monoamine neurotransmitters 1-methyl-4-phenylpyridinium and biogenic amines including serotonin, dopamine, norepinephrine and epinephrine. May play a role in regulating central nervous system homeostasis of monoamine neurotransmitters. May be involved in luminal transport of organic cations in the kidney and seems to use luminal proton gradient to drive organic cation reabsorption. Does not seem to transport nucleoside and nucleoside analogs such as uridine, cytidine, thymidine, adenosine, inosine, guanosine, and azidothymidine. In (PubMed:16873718) adenosine is efficiently transported but in a fashion highly sensitive to extracellular pH, with maximal activity in the pH range 5.5 to 6.5. Glu-206 is essential for the cation selectivity and may function as the charge sensor for cationic substrates. Transport is chloride and sodium-independent but appears to be sensitive to changes in membrane potential. Weakly inhibited by the classical inhibitors of equilibrative nucleoside transport, dipyridamole, dilazep, and nitrobenzylthioinosine. May play a role in the regulation of extracellular adenosine concentrations in cardiac tissues, in particular during ischemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC29A4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat