Anti-CYP11B1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-3898R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 11, Subfamily B, Polypeptide 1
- Antigen symbol:CYP11B1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1584
- Antigen synonyms:CPN1|P450C11|cytochrome P450|cytochrome P-450c11|cytochrome P450 11B1|subfamily XIB (steroid 11-beta-hydroxylase)|FHI|cytochrome p450 XIB1|polypeptide 1|cytochrome P450C11|mitochondrial|steroid 11-beta-monooxygenase|CYP11B|steroid 11-beta-hydroxylase|CYPXIB1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:350-400/503
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Has steroid 11-beta-hydroxylase activity. In addition to this activity, the 18 or 19-hydroxylation of steroids and the aromatization of androstendione to estrone have also been ascribed to cytochrome P45 XIB.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CYP11B1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: