Anti-GABRG2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-4112R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid (GABA) A Receptor gamma 2
- Antigen symbol:GABRG2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2566
- Antigen synonyms:CAE2|ECA2|GEFSP3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:30-80/475
- Purification:Purified by Protein A
- Pk:100 µl
Component of the heteropentameric receptor for GABA, the major inhibitory neurotransmitter in the vertebrate brain. Functions also as histamine receptor and mediates cellular responses to histamine. Functions as receptor for diazepines and various anesthetics, such as pentobarbital; these are bound at a separate allosteric effector binding site. Functions as ligand-gated chloride channel.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GABA A Receptor gamma 2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat