Order Entry
Slovenia
ContactUsLinkComponent
Anti-NUP210 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-NUP210 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13495R-FITC
Supplier:  Bioss
Anti-NUP210 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-13495R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nucleoporin 210kDa
  • Antigen symbol:
    NUP210
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    23225
  • Antigen synonyms:
    GP210|POM210
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

The nuclear pore complex (NPC) mediates bidirectional macromolecular traffic between the nucleus and cytoplasm in eukaryotic cells and is comprised of more than 100 different subunits. Many of the subunits belong to a family called nucleoporins (Nups), which are characterized by the presence of O-linked-N-acetylglucosamine moieties and a distinctive pentapeptide repeat (XFXFG). gp210, also known as Nup210 (nucleoporin 210kDa) or POM210, is a 1,887 amino acid single-pass type I membrane protein that localizes to both the endoplasmic reticulum and to the nucleus, specifically within the NPC. Expressed ubiquitously with highest expression in pancreas, testis, lung, ovary and liver, gp210 functions as a nucleoporin that is capable of dimerization and is essential for the assembly, fusion and structural integrity of the NPC. gp210 exists as multiple alternatively spliced isoforms and is subject to post-translational phosphorylation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NUP210
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat