Order Entry
Slovenia
ContactUsLinkComponent
Anti-DOCK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-DOCK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12432R-A488
Supplier:  Bioss
Anti-DOCK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-12432R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    dedicator of cytokinesis 4
  • Antigen symbol:
    DOCK4
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9732
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

DOCK 4 is a cytoplasmic peripheral membrane protein that belongs to the DOCK family of cytokinesis-regulating proteins. Expressed ubiquitously with highest expression in prostate, ovary and skeletal muscle, DOCK 4 functions as a guanine nucleotide exchange factor (GEF) that activates the small GTPase Rap 1 and, via this activation, plays a role in the regulation of adherens junctions between cells. Similar to other DOCK family members, DOCK 4 contains an N-terminal SH3 domain, a C-terminal proline-rich region and two internal DOCK homology regions designated DHR1 and DHR2. Defects in the gene encoding DOCK 4 result in the inactivation of Rap 1 and are, thus, implicated in the pathogenesis of various cancers such as ovarian, prostate, glioma and colorectal carcinomas. Four isoforms of DOCK 4 are expressed due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DOCK4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat