Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-ACVR2A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-12416R-A555
 :  Bioss
Anti-ACVR2A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-12416R-A555
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Activin A Receptor Type 2A
  • Antigen symbol:
    ACVR2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    92
  • Antigen synonyms:
    Activin receptor type IIA|Activin receptor type-2A [Precursor]|ACTR-IIA|ACVR2|ACTRIIA
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Members of the transforming growth factor b superfamily bind to a pair of transmembrane proteins, known as receptor types I and II, which contain serine/threonine kinases and associate to form a signaling complex (1). Activin has been shown to bind a heteromeric noncovalent complex, which consists of a type I receptor, ACTR-IA (also designated ACVRI and ALK-2) or ACTR-IB (also designated ALK-4 and SKR2), and a type II receptor, ACTR-IIA (also designated ACVR2A) or ACTR-IIB (also designated ACVR2B) (1-6). Both receptor types are highly expressed in brain (5). The activin receptor family members are thought to mediate distinct effects on gene expression, cell differentiation, and morpho- genesis in a dose dependent fashion (5,6).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACVR2A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat