- Antibody type:Primary
- Antigen name:dedicator of cytokinesis 5
- Antigen symbol:DOCK5
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:80005
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
DOCK 5 is a 1,870 amino acid protein belonging to the DOCK family of cytokinesis-regulating proteins. This cytoplasmic peripheral membrane protein activates Rac 1 and Rac 2 small GTPases, while presumably acting as a guanine nucleotide exchange factor (GEF), which exchanges bound GDP for free GTP. DOCK 5 contains one DHR-1 (CZH-1) domain, one DHR-2 (CZH-2) domain and one SH3 domain. The DHR-2 domain is a putative GEF activity mediator. In mice, spontaneous mutation of the gene encoding DOCK 5 leads to deletion of the DHR-1 domain, which functions to bind phospholipids and assists in protein-protein interactions, resulting in rupture of lens cataract (RLC). Due to siRNA knockdown studies, it is suspected that DOCK 5 may also be an important mediator of CrkII/CrkL regulation of Caco-2 migration and spreading on COL4. There are two isoforms of DOCK 5 that exist as a result of alternative splicing events.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: DOCK5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat