Anti-ADAMTS1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-1208R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase with Thrombospondin Type 1 Motif, 1
- Antigen symbol:ADAMTS1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:11504
- Antigen synonyms:ADAMTS|ADAM-TS1|METH1|ADAMTS-1|C3-C5|METH-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:920-967/967
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Cleaves aggrecan, a cartilage proteoglycan, and may be involved in its turnover. Has angiogenic inhibitor activity (By similarity). Active metalloprotease, which may be associated with various inflammatory processes as well as development of cancer cachexia. May play a critical role in follicular rupture (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ADAMTS1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat