Specifications
- Antibody type:Primary
- Antigen name:Aldehyde dehydrogenase 3 family member A2
- Antigen symbol:ALDH3A2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ALDH10|aldehyde dehydrogenase 3A2|DKFZp686E23276|FLJ20851|fatty aldehyde dehydrogenase|aldehyde dehydrogenase 10|FALDH|SLS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Aldehyde dehydrogenases (ALDHs) mediate the NADP+-dependent oxidation of aldehydes into acids and play an important role in the detoxification of alcohol-derived acetaldehyde, as well as in lipid peroxidation and in the metabolism of corticosteroids, biogenic amines and neurotransmitters. ALDH3A2 (aldehyde dehydrogenase 3 family, member A2), also known as SLS, FALDH or ALDH10, is a 485 amino acid single-pass membrane protein that localizes to the cytoplasmic side of the endoplasmic reticulum and belongs to the aldehyde dehydrogenase family. Expressed in a variety of tissues, including liver, heart, lung, brain, kidney and placenta, ALDH3A2 catalyzes the NAD+-dependent oxidation of long-chain aliphatic aldehydes to fatty acids, a process that is necessary for detoxification and lipid metabolism. Defects in the gene encoding ALDH3A2 are the cause of Sjoegren-Larsson syndrome (SLS), an autosomal recessive neurocutaneous disorder characterized by severe mental retardation, seizures and speech defects. Multiple isoforms of ALDH3A2 exist due to alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ALDH3A2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat