Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubing
Dialysis Tubing
Dialysis tubing
Dialysis tubing
Dialysis tubing
Catalog # KLEI1214355
Supplier:
KLEINFELD
Expand image
Dialysis tubing
Catalog # KLEI1214355
Supplier:
KLEINFELD
Specifications
Ø:
14,3 mm
MWCO:
0,012 - 14 kD
Pk:
30 m
Specifications
About this item
Product Details
Product Details