Anti-p-WNK1 Rabbit Polyclonal Antibody [clone: T60]
Catalog # BWRLBS4208
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-WNK1
- Clonality:Polyclonal
- Clone:T60
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human WNK1 around the phosphorylation site of Threonine 60.
p-WNK1 (T60) pAb detects endogenous levels of WNK1 protein only when phosphorylated at Thr60.
Type: Primary
Antigen: p-WNK1
Clonality: Polyclonal
Clone: T60
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat