Anti-IFN-gammaRalpha Rabbit Polyclonal Antibody [clone: G456]
Catalog # BWRLBS1667
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IFN-gammaR alpha
- Clonality:Polyclonal
- Clone:G456
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 430-480 of Human IFN-gammaRalpha.
IFN-?R? (G456) pAb detects endogenous levels of IFN-?R? protein.
Type: Primary
Antigen: IFN-gammaRalpha
Clonality: Polyclonal
Clone: G456
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat