Anti-Tetraspanin-8 Rabbit Polyclonal Antibody [clone: S129]
BWRLBS2467
- Antibody type:Primary
- Antigen symbol:Tetraspanin-8
- Clonality:Polyclonal
- Clone:S129
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 110-160 of Human Tetraspanin-8.
Tetraspanin-8 (S129) pAb detects endogenous levels of Tetraspanin-8 protein.
Type: Primary
Antigen: Tetraspanin-8
Clonality: Polyclonal
Clone: S129
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human