Anti-PKC-pan Rabbit Polyclonal Antibody [clone: T495]
Catalog # BWRLBS1580
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PKC-pan
- Clonality:Polyclonal
- Clone:T495
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 470-520 of Human PKC-pan.
PKC-pan (T495) pAb detects endogenous levels of PKC ?, ? I, ? II, ?, ?, ?, ?, ? and ? isoforms.
Type: Primary
Antigen: PKC-pan
Clonality: Polyclonal
Clone: T495
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat