Anti-CHOP Rabbit Polyclonal Antibody [clone: Q26]
Catalog # BWRLBS1527
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CHOP
- Clonality:Polyclonal
- Clone:Q26
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids N-terminus of Human CHOP.
CHOP (Q26) pAb detects endogenous levels of CHOP protein.
Type: Primary
Antigen: CHOP
Clonality: Polyclonal
Clone: Q26
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse