Anti-C5aR Rabbit Polyclonal Antibody [clone: S334]
BWRLBS1522
- Antibody type:Primary
- Antigen symbol:C5aR
- Clonality:Polyclonal
- Clone:S334
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 300-350 of Human C5aR.
CD88/C5aR (S334) pAb detects endogenous levels of D88/C5aR protein.
Type: Primary
Antigen: C5aR
Clonality: Polyclonal
Clone: S334
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat