Anti-VASP Rabbit Polyclonal Antibody [clone: H150]
BWRLBS1433
- Antibody type:Primary
- Antigen symbol:VASP
- Clonality:Polyclonal
- Clone:H150
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 130-180 of Human VASP.
VASP (H150) pAb detects endogenous levels of VASP protein.
Type: Primary
Antigen: VASP
Clonality: Polyclonal
Clone: H150
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat