Anti-UMP-CMP Kinase Rabbit Polyclonal Antibody [clone: E36]
Catalog # BWRLBS1975
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:UMP-CMP Kinase
- Clonality:Polyclonal
- Clone:E36
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human UMP-CMP Kinase.
UMP-CMP Kinase (E36) pAb detects endogenous levels of UMP-CMP Kinase protein.
Type: Primary
Antigen: UMP-CMP Kinase
Clonality: Polyclonal
Clone: E36
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat