Anti-c-TAK1 Rabbit Polyclonal Antibody [clone: P8]
Catalog # BWRLBS1969
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:c-TAK1
- Clonality:Polyclonal
- Clone:P8
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human c-TAK1.
c-TAK1 (P8) pAb detects endogenous levels of C-TAK1 protein.
Type: Primary
Antigen: c-TAK1
Clonality: Polyclonal
Clone: P8
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat