Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Hyperosid (from Hypericum perforatum) ≥97.0% (by HPLC), pure

100 mg
Catalog # A1791.0100
Irritant
Supplier:   PanReac AppliChem
Hyperosid (from Hypericum perforatum) ≥97.0% (by HPLC), pure
Catalog # A1791.0100
Supplier:  PanReac AppliChem
CAS Number:  482-36-0
Hyperosid (from Hypericum perforatum) ≥97.0% (by HPLC), pure

100 mg

CAS Number:  482-36-0
Price
633,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Originated from hypericum perforatum.

Synonyms:  Hyperoside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, Hyperin, Quercetin-3-galactoside
  • CAS Number:482-36-0
  • Formula:C₂₁H₂₀O₁₂
  • MDL Number:MFCD00016933
  • Molecular Weight:464.38 g/mol
  • Storage Temperature:Refrigerator

 

Specifications

Specification Test Results

  • State of Matter:
    Solid
  • Storage Temperature:
    2 - 8 °C
  • Assay (HPLC):
    min. 97%

Specifications

  • Pk:
    100 mg

Specifications