
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, white powder for electrophoresis
Catalog # ICNA04856126
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:≥99.5%
- FTIR:Conforms to Standard
Specification Test Results
About this item
:
:154.25 g/mol
Melting Pt:40…50 °C
Storage Temperature:Refrigerator
UN:3335
ADR:9,III
:GHS07
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877