Order Entry
Slovenia
ContactUsLinkComponent
Aspergillus niger Type X glucose oxidase, MP Biomedicals
Aspergillus niger Type X glucose oxidase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Aspergillus niger Type X glucose oxidase, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    9001-37-0
  • Species:
    Aspergillus niger Type X
  • Storage conditions:
    −20 °C
  • Form:
    Yellow lyophilized powder
  • Enzyme name:
    Glucose oxidase
  • Enzyme activity:
    >100 u/mg material
  • Molecular weight:
    ~160,000 (Lit.)
  • pH:
    pH Optimum: 6.5, pH Stability: 8.0, pH Range: 4 - 7

Specifications

Frequently Bought Together