Tetrakisacetonitrile copper(I) triflate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:58452-28-1
:
Formula:Cu(NCCH₃)₄·CF₃SO₃
MDL Number:MFCD10566992
:376.821 g/mol
Melting Pt:29…33 °C
Density:1.294 (25 °C)
UN:1759
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS05
Specification Test Results
- Appearance (Color):White
- Appearance (Form):Powder, Crystalline Powder, Crystals, and/or Chunks
- Infrared spectrum:Conforms to Structure
- Complexometric Titration (CU):16,18 - 17,53%
- Fluorine NMR Spectra:Conforms to Structure
- Proton NMR spectra:Conforms to Structure
Specification Test Results
Frequently Bought Together