Desalting column, G-Trap™ GT
Supplier: G-Biosciences
Certificates
About this item
G-Trap™ GT desalting columns are prepacked ready to use columns for separation of low molecular weight substances from high molecular weight substances based on size exclusion chromatography. Mostly used for removal of salt or buffer exchange before or after different chromatographic steps. They can also be used for separation of proteins based on their sizes.
- Made of biocompatible PP, which does not interact with biomolecules
- Stopper at the inlet and snap-off end at the outlet
The G-Trap™ GT desalting columns can be operated with syringe, peristaltic pump or a chromatography system.
Delivery: Supplied with one ¹/₁₆" male/Luer female union for connecting a syringe to a G-Trap™ GT desalting column and two female ¹/₁₆" stop plugs for sealing the bottom of a G-Trap™ GT desalting column.
Specifications
- Catalog No:
- GENO786-1024
- GENO786-1025
- GENO786-1028
- GENO786-1023
- GENO786-1026
- GENO786-1027
- Max. pressure:
- 5 bar
- 5 bar
- 5 bar
- 5 bar
- 5 bar
- 5 bar
- pH range:
- 2 - 13
- 2 - 13
- 2 - 13
- 2 - 13
- 2 - 13
- 2 - 13