Chemical and cold resistant gloves, Tricotril® Winter 738
Supplier: KCL by Honeywell
Certificates
About this item
Tricotril® Winter 738 is made in a special manufacturing process and consists of a sealed nitrile layer on a wool liner. This allows efficient working in cold environments (down to −20 °C) or with cold objects. The glove also offers good heat protection up to +100 °C.
- Warm seamless wool liner with skin friendly, solvent-free adhesive
- Wool liner is fixed later to the finished glove, to ensure maximum protection
- Good antistatic and electrostatic properties
- Short-term cold protection down to −50 °C
- Broad chemical resistance, high mechanical resistance
- Good grip when handling greasy and oily parts
- Free from natural latex
Typical applications include: Chemical industries, maintenance, food industries, industrial cleaning, petro-chemicals, working in cold areas, working in areas with momentary steam hazards, handling of contaminated heavy parts, and galvanising works.
Certifications: EN ISO 3741- Type A (AJKLOPT), EN 388 (3121X), EN 407 (X1XXXX); Cat. III; AQL <0,65
Food contact approved
Food contact approved
Specifications
- Catalog No:
- KCHL738.11
- KCHL738.9
- KCHL738.10
- KCHL738.8
- Model:
- 738
- 738
- 738
- 738
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Wool
- Wool
- Wool
- Wool
- Finish:
- Textured palm
- Textured palm
- Textured palm
- Textured palm
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN ISO 3741- Type A (AJKLOPT), EN 388 (3121X), EN 407 (X1XXXX)
- EN ISO 3741- Type A (AJKLOPT), EN 388 (3121X), EN 407 (X1XXXX)
- EN ISO 3741- Type A (AJKLOPT), EN 388 (3121X), EN 407 (X1XXXX)
- EN ISO 3741- Type A (AJKLOPT), EN 388 (3121X), EN 407 (X1XXXX)
- Food contact approved:
- +
- +
- +
- +
Specifications
Frequently Bought Together